  • Search Results
  • Deletion or inhibition of Fc gamma receptor 2B (CD32) prevents FVIII-specific activation of memory B cells in vitro.
Deletion or inhibition of Fc gamma receptor 2B (CD32) prevents FVIII-specific activation of memory B cells in vitro.
Thrombosis and haemostasis (2015-08-08)
Sonja Werwitzke, Nadine Vollack, Marcus von Hornung, Katy Kalippke, Julia Kutzschbach, Arne Trummer, Arnold Ganser, Andreas Tiede
ABSTRACT
Development of inhibitory antibodies against factor VIII (FVIII) is a severe complication of replacement therapy in haemophilia A. Patients with inhibitors are treated with high FVIII doses in the context of immune tolerance therapy (ITT). Data from haemophilia A mouse model suggest that high FVIII concentrations prevent the formation of antibody secreting cells (ASCs) from memory B cells (MBCs) by inducing apoptosis. Fc gamma receptor 2B (CD32) is an important regulator of B cell function, mediating inhibitory signals after cross-linking with the B cell receptor. Here, the role of CD32 in the regulation of FVIII-specific MBCs was investigated using F8-/- and F8-/-CD32-/- knockout mice and monoclonal antibodies (mAbs). The initial immune response was similar between F8-/- and F8-/-CD32-/- mice, including concentration of anti-FVIII antibodies and number of FVIII-specific ASCs in spleen and bone marrow. In contrast, formation of ASCs from MBCs upon rhFVIII re-stimulation in vitro was abolished in F8-/-CD32-/- mice, whereas FVIII/anti-FVIII immune complexes significantly enhanced ASC formation in F8-/- mice. Inhibition of CD32 by mAbs or F(ab)2 fragments prevented ASC formation in a dose-dependent manner. Transfer of B cell-depleted splenocytes using CD45R (B220) depletion from CD32-competent mice did not restore ASC formation in F8-/-CD32-/- cells confirming that CD32 is required on B cells. We conclude that CD32 is a crucial regulator of FVIII-specific B cells and is required for the differentiation of MBCs into ASCs. Inhibition of CD32 could potentially improve the efficacy of FVIII in the context of ITT.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing