  • Search Results
  • Ischemia-reperfusion rat model of acute pancreatitis: protein carbonyl as a putative early biomarker of pancreatic injury.
Ischemia-reperfusion rat model of acute pancreatitis: protein carbonyl as a putative early biomarker of pancreatic injury.
Clinical and experimental medicine (2014-06-18)
Alberto Schanaider, Thales Penna de Carvalho, Simone de Oliveira Coelho, Juan Miguel Renteria, Elis Cristina Araújo Eleuthério, Morgana Teixeira Lima Castelo-Branco, Kalil Madi, Wagner Baetas-da-Cruz, Heitor Siffert Pereira de Souza
ABSTRACT
Acute pancreatitis (AP) is an inflammatory disorder that can affect adjacent and/or remote organs. Some evidence indicates that the production of reactive oxygen species is able to induce AP. Protein carbonyl (PC) derivatives, which can also be generated through oxidative cleavage mechanisms, have been implicated in several diseases, but there is little or no information on this biomarker in AP. We investigated the association between some inflammatory mediators and PC, with the severity of ischemia-reperfusion AP. Wistar rats (n = 56) were randomly assigned in the following groups : control; sham, 15- or 180-min clamping of splenic artery, with 24 or 72 h of follow-up. The relationships between serum level of PC and thiobarbituric acid reactive species (TBARS) to myeloperoxidase (MPO) activity in tissue homogenates and to cytokines in culture supernatants of pancreatic samples were analyzed. MPO activity was related to the histology scores and increased in all clamping groups. Tumor necrosis factor-alpha (TNF-α), interleukin 1 beta (IL-1β), and interleukin-6 were higher in the 180-min groups. Significant correlations were found between MPO activity and the concentrations of TNF-α and IL-1β. PC levels increased in the 15-min to 24-h group. TBARS levels were not altered substantially. MPO activity and TNF-α and IL-1β concentrations in pancreatic tissue are correlated with AP severity. Serum levels of PC appear to begin to rise early in the course of the ischemia-reperfusion AP and are no longer detected at later stages in the absence of severe pancreatitis. These data suggest that PC can be an efficient tool for the diagnosis of early stages of AP.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
Sigma-Aldrich
o-Dianisidine, peroxidase substrate
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
SAFC
Hexadecyltrimethylammonium bromide