  • Search Results
  • Association between Stress Response Genes and Features of Diurnal Cortisol Curves in the Multi-Ethnic Study of Atherosclerosis: A New Multi-Phenotype Approach for Gene-Based Association Tests.
Association between Stress Response Genes and Features of Diurnal Cortisol Curves in the Multi-Ethnic Study of Atherosclerosis: A New Multi-Phenotype Approach for Gene-Based Association Tests.
PloS one (2015-05-21)
Zihuai He, Erin K Payne, Bhramar Mukherjee, Seunggeun Lee, Jennifer A Smith, Erin B Ware, Brisa N Sánchez, Teresa E Seeman, Sharon L R Kardia, Ana V Diez Roux
ABSTRACT
The hormone cortisol is likely to be a key mediator of the stress response that influences multiple physiologic systems that are involved in common chronic disease, including the cardiovascular system, the immune system, and metabolism. In this paper, a candidate gene approach was used to investigate genetic contributions to variability in multiple correlated features of the daily cortisol profile in a sample of European Americans, African Americans, and Hispanic Americans from the Multi-Ethnic Study of Atherosclerosis (MESA). We proposed and applied a new gene-level multiple-phenotype analysis and carried out a meta-analysis to combine the ethnicity specific results. This new analysis, instead of a more routine single marker-single phenotype approach identified a significant association between one gene (ADRB2) and cortisol features (meta-analysis p-value=0.0025), which was not identified by three other commonly used existing analytic strategies: 1. Single marker association tests involving each single cortisol feature separately; 2. Single marker association tests jointly testing for multiple cortisol features; 3. Gene-level association tests separately carried out for each single cortisol feature. The analytic strategies presented consider different hypotheses regarding genotype-phenotype association and imply different costs of multiple testing. The proposed gene-level analysis integrating multiple cortisol features across multiple ethnic groups provides new insights into the gene-cortisol association.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications