  • Search Results
  • Myelinated mouse nerves studied by X-ray phase contrast zoom tomography.
Myelinated mouse nerves studied by X-ray phase contrast zoom tomography.
Journal of structural biology (2015-11-08)
M Bartels, M Krenkel, P Cloetens, W Möbius, T Salditt
ABSTRACT
We have used X-ray phase contrast tomography to resolve the structure of uncut, entire myelinated optic, saphenous and sciatic mouse nerves. Intrinsic electron density contrast suffices to identify axonal structures. Specific myelin labeling by an osmium tetroxide stain enables distinction between axon and surrounding myelin sheath. Utilization of spherical wave illumination enables zooming capabilities which enable imaging of entire sciatic internodes as well as identification of sub-structures such as nodes of Ranvier and Schmidt-Lanterman incisures.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Tannic acid, ACS reagent
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Tannic acid
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Tannic acid, puriss., meets analytical specification of USP, powder
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Tannic acid, Source: Chinese natural gall nuts
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC