  • Search Results
  • Aspergillus Associated with Meju, a Fermented Soybean Starting Material for Traditional Soy Sauce and Soybean Paste in Korea.
Aspergillus Associated with Meju, a Fermented Soybean Starting Material for Traditional Soy Sauce and Soybean Paste in Korea.
Mycobiology (2015-11-06)
Seung-Beom Hong, Dae-Ho Kim, Robert A Samson
ABSTRACT
Aspergillus is an important fungal genus used for the fermentation of Asian foods; this genus is referred to as koji mold in Japan and China. A. oryzae, A. sojae, and A. tamari are used in the production of miso and shoyu in Japan, but a comprehensive taxonomic study of Aspergillus isolated from Meju, a fermented soybean starting material for traditional soy sauce and soybean paste in Korea, has not been conducted. In this study, various Aspergillus species were isolated during a study of the mycobiota of Meju, and the aspergilli were identified based on phenotypic characteristics and sequencing of the β-tubulin gene. Most strains of Aspergillus were found to belong to the following sections: Aspergillus (n = 220), Flavi (n = 213), and Nigri (n = 54). The most commonly identified species were A. oryzae (n = 183), A. pseudoglaucus (Eurotium repens) (n = 81), A. chevalieri (E. chevalieri) (n = 62), A. montevidensis (E. amstelodami) (n = 34), A. niger (n = 21), A. tamari (n = 15), A. ruber (E. rubrum) (n = 15), A. proliferans (n = 14), and A. luchuensis (n = 14); 25 species were identified from 533 Aspergillus strains. Aspergillus strains were mainly found during the high temperature fermentation period in the later steps of Meju fermentation.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)