  • Search Results
  • Histone methyltransferase SETDB1 regulates liver cancer cell growth through methylation of p53.
Histone methyltransferase SETDB1 regulates liver cancer cell growth through methylation of p53.
Nature communications (2015-10-17)
Qi Fei, Ke Shang, Jianhua Zhang, Shannon Chuai, Desheng Kong, Tianlun Zhou, Shijun Fu, Ying Liang, Chong Li, Zhi Chen, Yuan Zhao, Zhengtian Yu, Zheng Huang, Min Hu, Haiyan Ying, Zhui Chen, Yun Zhang, Feng Xing, Jidong Zhu, Haiyan Xu, Kehao Zhao, Chris Lu, Peter Atadja, Zhi-Xiong Xiao, En Li, Jianyong Shou
ABSTRACT
SETDB1 is a histone H3K9 methyltransferase that has a critical role in early development. It is located within a melanoma susceptibility locus and facilitates melanoma formation. However, the mechanism by which SETDB1 regulates tumorigenesis remains unknown. Here we report the molecular interplay between SETDB1 and the well-known hotspot gain-of-function (GOF) TP53 R249S mutation. We show that in hepatocellular carcinoma (HCC) SETDB1 is overexpressed with moderate copy number gain, and GOF TP53 mutations including R249S associate with this overexpression. Inactivation of SETDB1 in HCC cell lines bearing the R249S mutation suppresses cell growth. The TP53 mutation status renders cancer cells dependent on SETDB1. Moreover, SETDB1 forms a complex with p53 and catalyses p53K370 di-methylation. SETDB1 attenuation reduces the p53K370me2 level, which subsequently leads to increased recognition and degradation of p53 by MDM2. Together, we provide both genetic and biochemical evidence for a mechanism by which SETDB1 regulates cancer cell growth via methylation of p53.
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥48%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Hydrofluoric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥40%
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53