Suppression of FoxO6 by lipopolysaccharide in aged rat liver.
Oncotarget (2015-10-28)
Dae Hyun Kim, Min Hi Park, Ki Wung Chung, Min Jo Kim, Daeui Park, Bonggi Lee, Eun Kyeong Lee, Yeon Ja Choi, Nam Deuk Kim, Byung Pal Yu, Hae Young Chung
ABSTRACT
The beneficial role of FoxO during aging has been proposed for its promotion of resistance to oxidative stress and inhibition of pro-inflammatory mediators. On the other hand, NF-κB is a pro-inflammatory transcription factor which is a key mediator of inflammatory cytokine generation. However, the correlation between FoxO6 and NF-κB during aging has not fully been explored.The main purpose of the present study was to elucidate mechanisms underlying the protective role of FoxO6 in the maintenance of cellular homeostasis under potent pro-inflammatory conditions induced by LPS. Initial experimentation revealed that reduced FoxO6 activity during aging was caused by its phosphorylation, which suppressed its transcriptional activity in aged livers. Transfection with FoxO6-wt virus and FoxO6-siRNA in HepG2 cells revealed that FoxO6 phosphorylation by LPS leads to NF-κB activation via Akt and Pak1 pathways. Furthermore, Pak1 activity was increased in a phosphatidylinositol 3-kinase independent manner, and LPS-induced FoxO6 phosphorylation and FoxO6 inactivation were Pak1-dependent in nuclear fractions of cells. Further revealed Pak1 phosphorylation by LPS permitted interaction between FoxO6 and Akt.Current study suggests FoxO6 phosphorylation facilitates the nuclear translocation of NF-κB via Akt and Pak1 pathways induced by LPS in aged rats.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems