  • Search Results
  • Use of cloud-point preconcentration for spectrophotometric determination of trace amounts of antimony in biological and environmental samples.
Use of cloud-point preconcentration for spectrophotometric determination of trace amounts of antimony in biological and environmental samples.
Analytical biochemistry (2015-08-19)
Abdel-Azeem M El-Sharjawy, Alaa S Amin
ABSTRACT
This work presents a cloud-point extraction process using the micelle-mediated extraction method for simultaneous preconcentration and determination of Sb(III) and Sb(V) species in biological and environmental samples as a prior preconcentration step to their spectrophotometric determination. The analytical system is based on the selective reaction between Sb(III) and 3-dichloro-6-(3-carboxy-2-hydroxy-1-naphthylazo)quinoxaline (DCHNAQ) in the presence of cetyltrimethylammonium bromide (CTAB) and potassium iodide at pH 4.5. Total Sb concentration was determined after reduction of Sb(V) to Sb(III) in the presence of potassium iodide and ascorbic acid. The optimal reaction conditions and extraction were studied, and the analytical characteristics of the method (e.g., limits of detection and quantification, linear range, preconcentration, improvement factors) were obtained. Linearity for Sb(III) was obeyed in the range of 0.2-20 ng ml(-1). The detection and quantification limits for the determination of Sb(III) were 0.055 and 0.185 ng ml(-1), respectively. The method has a lower detection limit and wider linear range, inexpensive instrument, and low cost, and is more sensitive compared with most other methods. The interference effect of some anions and cations was also studied. The method was applied to the determination of Sb(III) in the presence of Sb(V) and total antimony in blood plasma, urine, biological, and water samples.
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
SAFC
Hexadecyltrimethylammonium bromide
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis