  • Search Results
  • In silico and biological analysis of anti-androgen activity of the brominated flame retardants ATE, BATE and DPTE in zebrafish.
In silico and biological analysis of anti-androgen activity of the brominated flame retardants ATE, BATE and DPTE in zebrafish.
Chemico-biological interactions (2015-03-31)
Ajay Pradhan, Solomon Asnake, Joubert Banjop Kharlyngdoh, Carina Modig, Per-Erik Olsson
ABSTRACT
The brominated flame retardants (BFRs) 1,2-dibromo-4-(1,2-dibromoethyl)cyclohexane (TBECH or DBE-DCBH) and allyl 2,4,6-tribromophenyl ether (ATE or TBP-AE) are alternative BFRs that have been introduced to replace banned BFRs. TBECH is a potential endocrine disrupter in human, chicken and zebrafish and in a recent study we showed that ATE, along with the structurally similar BFR 2,3-dibromopropyl 2,4,6-tribromophenyl ether (DPTE or TBP-DBPE) and its metabolite 2-bromoallyl 2,4,6-tribromophenyl ether (BATE or TBP-BAE) are potential endocrine and neuronal disrupters in human. In this study we analyzed ATE, BATE and DPTE for zebrafish androgen receptor (zAR) modulating properties. In silico analysis with two softwares, Molecular Operating Environment (MOE) and Internal Coordinate Mechanics (ICM), showed that ATE, BATE and DPTE bind to zAR. In vitro AR activation assay revealed that these three BFRs down-regulate 11-ketotestosterone (KT) mediated zAR activation. Exposure to 10 μM DPTE resulted in reduced hatching success and like TBECH, BATE and DPTE at 10 μM also had teratogenic properties with 20% and 50% back-bone curvature respectively. Gene transcription analysis in zebrafish embryos as well as in juveniles showed down-regulation of the androgen receptor and androgen response genes, which further support that these BFRs are androgen antagonists and potential endocrine disrupting compounds. Genes involved in steroidogenesis were also down-regulated by these BFRs. In view of this, the impact of these BFRs on humans and wildlife needs further analysis.
Sigma-Aldrich
11-Ketotestosterone
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phenylacetic acid, suitable for plant cell culture
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)