  • Search Results
  • The metal-organic framework HKUST-1 as efficient sorbent in a vortex-assisted dispersive micro solid-phase extraction of parabens from environmental waters, cosmetic creams, and human urine.
The metal-organic framework HKUST-1 as efficient sorbent in a vortex-assisted dispersive micro solid-phase extraction of parabens from environmental waters, cosmetic creams, and human urine.
Talanta (2015-04-18)
Priscilla Rocío-Bautista, Carla Martínez-Benito, Verónica Pino, Jorge Pasán, Juan H Ayala, Catalina Ruiz-Pérez, Ana M Afonso
ABSTRACT
Three metal-organic frameworks (MOFs), specifically HKUST-1, MOF-5, and MIL-53(Al), have been synthetized, characterized, studied and compared in a vortex-assisted dispersive micro-solid-phase extraction (VA-D-µ-SPE) procedure in combination with high-performance liquid chromatography (HPLC) with diode-array detection (DAD) for determining seven parabens in environmental waters (tap water, swimming pool water, and water coming from a spa pool), human urine (from two volunteers), and cosmetic creams (two commercial brands). Experimental parameters, such as nature and amount of MOF, sample volume, nature of elution solvent and its amount, vortex and centrifugation time, among others, were properly optimized. HKUST-1 was the most adequate MOF to work with. Detection limits for the overall method down to 0.1 μgL(-1) for butylparaben (BPB) and benzylparaben (BzPB) were obtained, with determination coefficients (R(2)) higher than 0.9966 for a range of 0.5-147 μgL(-1) (depending on the paraben), average relative recoveries (RR, in %) of 80.3% at the low spiked level (7 μgL(-1)), and relative standard deviation (RSD) values below 10% also at the low spiked level. The strength of the affinity between HKUST-1 and parabens was evaluated, and it ranged from 33.5% for isopropylparaben (iPPB) to 77.0% for isobutylparaben (iBPB). When analyzing complex environmental waters, RR values of 78%, inter-day precision values (as RSD) lower than 15%, and intra-day precision values lower than 7.8% were obtained, despite the observed matrix effect. When analyzing cosmetic creams, parabens were detected, with contents ranging from 0.14 ± 0.01 μgg(-1) for EPB in the healing cream analyzed to 1.12 ± 0.07 mgg(-1) for MPB in the mask cream analyzed, with precision values (RSD) lower than 12% and RR values from 63.7% for propylparaben (PPB) to 121% for iPPB. When analyzing human urine, no parabens were detected but the method could be performed with RSD values lower than 19%. These results show the adequateness of MOFs as sorbents in VA-D-µ-SPE procedures despite sample complexity.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
Sodium chloride solution, 5 M