  • Search Results
  • The OB-fold domain 1 of human POT1 recognizes both telomeric and non-telomeric DNA motifs.
The OB-fold domain 1 of human POT1 recognizes both telomeric and non-telomeric DNA motifs.
Biochimie (2015-05-03)
Kyung H Choi, Amanda S Lakamp-Hawley, Carol Kolar, Ying Yan, Gloria E O Borgstahl, Michel M Ouellette
ABSTRACT
The POT1 protein plays a critical role in telomere protection and telomerase regulation. POT1 binds single-stranded 5'-TTAGGGTTAG-3' and forms a dimer with the TPP1 protein. The dimer is recruited to telomeres, either directly or as part of the Shelterin complex. Human POT1 contains two Oligonucleotide/Oligosaccharide Binding (OB) fold domains, OB1 and OB2, which make physical contact with the DNA. OB1 recognizes 5'-TTAGGG whereas OB2 binds to the downstream TTAG-3'. Studies of POT1 proteins from other species have shown that some of these proteins are able to recognize a broader variety of DNA ligands than expected. To explore this possibility in humans, we have used SELEX to reexamine the sequence-specificity of the protein. Using human POT1 as a selection matrix, high-affinity DNA ligands were selected from a pool of randomized single-stranded oligonucleotides. After six successive rounds of selection, two classes of high-affinity targets were obtained. The first class was composed of oligonucleotides containing a cognate POT1 binding sites (5'-TTAGGGTTAG-3'). The second and more abundant class was made of molecules that carried a novel non-telomeric consensus: 5'-TNCANNAGKKKTTAGG-3' (where K = G/T and N = any base). Binding studies showed that these non-telomeric sites were made of an OB1-binding motif (TTAGG) and a non-telomeric motif (NT motif), with the two motifs recognized by distinct regions of the OB1 domain. POT1 interacted with these non-telomeric binding sites with high affinity and specificity, even when bound to its dimerization partner TPP1. This intrinsic ability of POT1 to recognize NT motifs raises the possibility that the protein may fulfill additional functions at certain non-telomeric locations of the genome, in perhaps gene transcription, replication, or repair.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
BICINE, ≥99% (titration)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
BICINE, BioXtra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%