  • Search Results
  • A Comparative In Vitro Study on the Antioxidant and Anti-acetylcholinesterase Properties of Aerial Parts of Strophanthus preusii Engl & Pax.
A Comparative In Vitro Study on the Antioxidant and Anti-acetylcholinesterase Properties of Aerial Parts of Strophanthus preusii Engl & Pax.
The West Indian medical journal (2015-03-18)
O A Adaramoye, A Olajuyin
ABSTRACT
To evaluate and compare the antioxidant and acetylcholinestrase (AChE)-inhibitory properties of aerial parts of Strophanthus preussii (leaves, stem and root named as SPL, SPS and SPR, respectively) while catechin served as standard. The antioxidant and AchE-inhibitory properties of the methanol extracts of SP were evaluated by standard in vitro methods viz: DPPH (2,2-diphenyl-1-picrylhydrazine), nitric oxide (NO), hydroxyl radical (OH-) and hydrogen peroxide (H2O2) radical scavenging assays as well as reducing power, Fe2+/ascorbate-induced lipid peroxidation (LPO) and AChE inhibition assays. The total phenolic and flavonoid contents of the extracts were also estimated. High phenolic and flavonoid contents were found in the aerial parts of Strophanthus preussii. The amount of phenolic and flavonoids contents followed the order SPL > SPR > SPS at 250─1000 µg/ml. The results revealed that all the extracts showed antioxidant activities in vitro. However, SPL had the highest DPPH, H2O2 and OH radical scavenging abilitie, while the reducing power of the extracts followed the order SPR > SPL > SPS at 1000 µg/ml. In addition, SPL, SPS and SPR significantly inhibited LPO in rat liver by 42%, 23%, 35% and in rat brain by 68%, 31% and 51%, respectively. The LPO inhibitory activities of SPL were statistically similar to the standard. Only SPS produced significant NO scavenging effects among the extracts. The percentage inhibition of AChE activity was significant for SPL and SPR at 750 and 1000 µg/ml. The leaves and root of Strophanthus preusii proved to be potent natural antioxidants and could justify their traditional use in the management of stress-related diseases.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
2-Deoxy-D-ribose, ≥99.0% (TLC)
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
2-Deoxy-D-ribose, 97%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Deoxy-D-ribose, suitable for cell culture, BioReagent
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%