  • Search Results
  • Antiplatelet aggregation and endothelial protection of I4, a new synthetic anti-diabetes sulfonylurea compound.
Antiplatelet aggregation and endothelial protection of I4, a new synthetic anti-diabetes sulfonylurea compound.
Platelets (2014-05-17)
Lingman Ma, Na Lu, Guanzhong Wu
ABSTRACT
I4 is a new synthetic anti-diabetes sulfonylurea compound. The aim of present study was to investigate the preventive effects and primary action mechanisms of I4 on platelet-mediated arterial thrombosis. Platelet aggregation and 5-hydroxytryptamine (5-HT) secretion ex vivo was detected. The time-to-occlusion (TTO), thrombus weight and content of von Willebrand factor (vWF) in rat model of electrical- and ferric chloride-induced vessel occlusion were determined. Meanwhile, a rat model of type 2 diabetes mellitus (T2DM) was established to evaluate the effect of I4 on levels of plasma p-selectin, 6-keto-prostaglandin F1a (6-keto-PGF1a), thromboxane B2 (TXB2), tissue-type plasminogen activator (t-PA) and type-1 plasminogen activator inhibitor (PAI-1). NO synthesis, NOS activity, adhesion of platelet toward endothelial cell and intercellular adhesion molecule-1 (ICAM-1) expression were examined. Results showed that I4 exhibited a higher inhibitory potency than Glimepiride on ADP-induced platelet aggregation and 5-HT release ex vivo. In addition, I4 reduced the thrombus weight and content of vWF and markedly prolonged TTO. Oral administration of I4 (1 ∼ 10 mg/kg) inhibited p-selectin production, elevated the ratio of plasma 6-keto-PGF1a/TXB2 and t-PA/PAI-1 in T2DM rats. Furthermore, I4 significantly improved NO synthesis and NOS activity, lowered adhesion ratio of platelet toward endothelial cells and ICAM-1 expression on HUVECs. These observations suggest that I4 markedly improves platelet-mediated arterial thrombosis by inhibiting platelet activation and release reaction, ameliorating the endothelial dysfunction such as the suppression of vWF production and the reduction of the overexpression of ICAM-1, displayed its potential in alleviating diabetes-associated vascular complications.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Phthaldialdehyde, for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder (may contain lumps)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Acetyl-5-hydroxytryptamine, ≥99% (TLC), powder
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)