  • Search Results
  • Randomized controlled trial of the prophylactic effect of urea-based cream on sorafenib-associated hand-foot skin reactions in patients with advanced hepatocellular carcinoma.
Randomized controlled trial of the prophylactic effect of urea-based cream on sorafenib-associated hand-foot skin reactions in patients with advanced hepatocellular carcinoma.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2015-02-11)
ZhengGang Ren, KangShun Zhu, HaiYan Kang, MinQiang Lu, ZengQiang Qu, LiGong Lu, TianQiang Song, WeiPing Zhou, Hui Wang, WeiZhu Yang, Xuan Wang, YongPing Yang, LeHua Shi, YuXian Bai, XiaoFeng Guo, Sheng-Long Ye
ABSTRACT
To assess whether urea-based cream (UBC) has prophylactic benefits on sorafenib-induced hand-foot skin reaction (HFSR) in patients with advanced hepatocellular carcinoma (HCC). In this randomized, open-label trial, 871 patients with advanced HCC throughout China were treated with 10% UBC three times per day plus best supportive care (BSC; n = 439) or BSC alone excluding all creams (n = 432), starting on day 1 of sorafenib treatment, for up to 12 weeks. HFSR was assessed every 2 weeks and at 14 weeks for patients completing the study. Once HFSR occurred, patients were allowed any cream, including a UBC. The 12-week incidence of any grade HFSR was significantly lower in the UBC group versus the BSC-alone group (56.0% v 73.6%, respectively; odds ratio [OR], 0.457; 95% CI, 0.344 to 0.608; P < .001), as was the incidence of grade ≥ 2 HFSR (20.7% v 29.2%, respectively; OR, 0.635; 95% CI, 0.466 to 0.866; P = .004). Median time to first occurrence of HFSR was significantly longer in the UBC group than the BSC-alone group (84 v 34 days, respectively; hazard ratio, 0.658; 95% CI, 0.541 to 0.799; P < .001). Elevated AST was associated with increased risk of HFSR but did not alter the treatment effect of UBC. UBC plus BSC, compared with BSC alone, did not affect the sorafenib dose reduction or interruption rate (9.1% v 11.8%, respectively; P = .1937), response rate (11.1% v 10.1%, respectively; P = .6674), or disease control rate (98.8% v 98.2%, respectively; P = .5350) at week 12. UBC prophylaxis in patients with advanced HCC starting sorafenib reduced HFSR rates, extended the time to first occurrence of HFSR, and improved patient quality of life compared with BSC. Blinded, randomized, placebo-controlled trials to determine the role of UBC on the incidence and severity of HFSR are warranted.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets