  • Search Results
  • A bioguided identification of the active compounds that contribute to the antiproliferative/cytotoxic effects of rosemary extract on colon cancer cells.
A bioguided identification of the active compounds that contribute to the antiproliferative/cytotoxic effects of rosemary extract on colon cancer cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-03-25)
Isabel Borrás-Linares, Almudena Pérez-Sánchez, Jesús Lozano-Sánchez, Enrique Barrajón-Catalán, David Arráez-Román, Alejandro Cifuentes, Vicente Micol, Antonio Segura Carretero
ABSTRACT
Rosemary extracts have exhibited potential cytostatic or cytotoxic effects in several cancer cell models but their bioactive compounds are yet to be discovered. In this work, the anticancer activity of a rosemary-leaf extract and its fractions were assayed to identify the phenolic compounds responsible for their antiproliferative/cytotoxic effects on a panel of human colon cancer cell lines. Bioguided fractionation of the rosemary-leaf extract was achieved by semi-preparative chromatography. The rosemary extract and the compounds in the fractions were characterized and quantified by HPLC-ESI-QTOF-MS. Cellular viability in the presence of these fractions and the whole extract was determined after 24 or 48 h incubations by using an MTT assay. Fractions containing diterpenes or triterpenes were the most active but not as much as the whole extract. In conclusion, carnosic acid, carnosol, 12-methoxycarnosic acid, taxodione, hinokione and betulinic acid were the putative candidates that contributed to the observed antiproliferative activity of rosemary in human colon cancer cells. Whether the effects of the extract and fractions are only cytostatic or cytotoxic needs to be elucidated. Nevertheless, the comparative antiproliferative study on the fractions and whole extract revealed potential synergistic effects between several components in the extract that may deserve further attention.
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Carnosic acid from Rosmarinus officinalis, ≥91%, powder
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid