  • Search Results
  • Prostate cancer cells stimulated by calcium-mediated activation of protein kinase C undergo a refractory period before re-releasing calcium-bearing microvesicles.
Prostate cancer cells stimulated by calcium-mediated activation of protein kinase C undergo a refractory period before re-releasing calcium-bearing microvesicles.
Biochemical and biophysical research communications (2015-03-24)
Dan Stratton, Colin Moore, Lei Zheng, Sigrun Lange, Jameel Inal
ABSTRACT
MVs are released in response to several stress agents, in an attempt to prevent continued cellular damage. After an initial stimulus of prostate cancer cells with sublytic C5b-9 and activation of MV release through PKC, cells take at least 20 min to fully recover their ability to microvesiculate. This release of MVs through activation of sublytic C5b-9 was inhibited by the PKC inhibitor bisindoylmaleimide I but not the Rho kinase inhibitor, Y27632. After stimulus there is a rise of 79 nMs(-1) over 11 s, reaching a peak [Ca(2+)]i of 920 nM. The concentration of cytosolic calcium then falls steadily at 2.4 nMs(-1) over 109 s reaching baseline levels (50-100 nM) within 10-15 min. In PC3 cells the rate of release of MVs from stimulated cells also reaches a minimum within 10-15 min. Using fura-2 AM-loaded cells, upon stimulation, cells were found to release MVs with a concentration of intravesicular calcium estimated at ∼ 430 nM.
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)