  • Search Results
  • Repression of Mammalian Target of Rapamycin Complex 1 Inhibits Intestinal Regeneration in Acute Inflammatory Bowel Disease Models.
Repression of Mammalian Target of Rapamycin Complex 1 Inhibits Intestinal Regeneration in Acute Inflammatory Bowel Disease Models.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-31)
Yuting Guan, Long Zhang, Xia Li, Xinyan Zhang, Shijie Liu, Na Gao, Liang Li, Ganglong Gao, Gaigai Wei, Zhaohua Chen, Yansen Zheng, Xueyun Ma, Stefan Siwko, Jin-Lian Chen, Mingyao Liu, Dali Li
ABSTRACT
The mammalian target of rapamycin (mTOR) signaling pathway integrates environmental cues to regulate cell growth and survival through various mechanisms. However, how mTORC1 responds to acute inflammatory signals to regulate bowel regeneration is still obscure. In this study, we investigated the role of mTORC1 in acute inflammatory bowel disease. Inhibition of mTORC1 activity by rapamycin treatment or haploinsufficiency of Rheb through genetic modification in mice impaired intestinal cell proliferation and induced cell apoptosis, leading to high mortality in dextran sodium sulfate- and 2,4,6-trinitrobenzene sulfonic acid-induced colitis models. Through bone marrow transplantation, we found that mTORC1 in nonhematopoietic cells played a major role in protecting mice from colitis. Reactivation of mTORC1 activity by amino acids had a positive therapeutic effect in mTORC1-deficient Rheb(+/-) mice. Mechanistically, mTORC1 mediated IL-6-induced Stat3 activation in intestinal epithelial cells to stimulate the expression of downstream targets essential for cell proliferation and tissue regeneration. Therefore, mTORC1 signaling critically protects against inflammatory bowel disease through modulation of inflammation-induced Stat3 activity. As mTORC1 is an important therapeutic target for multiple diseases, our findings will have important implications for the clinical usage of mTORC1 inhibitors in patients with acute inflammatory bowel disease.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis