One-pot synthesis of levulinic acid from cellulose in ionic liquids.
Bioresource technology (2015-06-10)
Yue Shen, Jian-Kui Sun, Yu-Xuan Yi, Bo Wang, Feng Xu, Run-Cang Sun
ABSTRACT
A simple and effective route for the production of levulinic acid (LA) from cellulose has been developed in SO3H-functionalized ionic liquids. The effects of ionic liquid structures, reaction conditions and combination of metal chlorides with ILs on the yield of LA were investigated, where the highest yield of 39.4% was obtained for 120 min in the presence of 1-(4-sulfonic acid) butyl-3-methylimidazolium hydrogen sulphate ([BSMim]HSO4) with addition of H2O. The catalytic activities of ionic liquids depended on the anions and decreased in the order: CF3SO3(-)>HSO4(-) > OAc(-), which was in good agreement with their acidity order. The ILs play a dual solvent-acid role for the cellulose conversion into LA and exhibited favorable catalytic activity over four repeated runs.
Sigma-Aldrich
D-(−)-Fructose, ≥99%
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Fructose, BioXtra
Sigma-Aldrich
D-(−)-Fructose, meets USP testing specifications
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C