  • Search Results
  • Designer SiO₂@Au nanoshells towards sensitive and selective detection of small molecules in laser desorption ionization mass spectrometry.
Designer SiO₂@Au nanoshells towards sensitive and selective detection of small molecules in laser desorption ionization mass spectrometry.
Nanomedicine : nanotechnology, biology, and medicine (2015-07-15)
Jinrui Gan, Xiang Wei, Yixin Li, Jiao Wu, Kun Qian, Baohong Liu
ABSTRACT
We report a novel platform using optimized SiO2@Au core-shell structures as matrices for highly efficient laser desorption/ionization mass spectrometry analysis of small biomolecules (MW<700 Da). Owing to the designer structure, SiO2@Au nanoshells can achieve low detection-of-limits (~pmol-fmol) in mass spectrometry and selective laser desorption/ionization in bio-mixtures towards diverse small molecules. By further surface modification with aptamers, Apt-SiO2@Au nanoshells allowed simultaneously targeted enrichment and detection of kanamycin with a detection limit at 200 pM. Our work not only starts new applications of SiO2@Au nanoshells in mass spectrometry, but also contributes to advanced analysis of either a group of small molecules or one target small molecule from complex bio-samples in a pre-designed manner for bio-diagnostics. Existing methods for the detection of small molecules are often not sensitive enough. Here, the authors developed aptamer functionalized SiO2@Au nanoshells for use in mass spectrometry, with very low detection limits. The new platform appeared to be simple and efficient and should be applicable in detection of clinical samples.
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
D-Mannitol, suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
DL-Phenylalanine, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%