  • Search Results
  • Combination of Genistein and Cisplatin with Two Designed Monofunctional Platinum Agents in Human Ovarian Tumour Models.
Combination of Genistein and Cisplatin with Two Designed Monofunctional Platinum Agents in Human Ovarian Tumour Models.
Anticancer research (2015-10-28)
Laila Arzuman, Philip Beale, Nick Proschogo, Jun Q Yu, Fazlul Huq
ABSTRACT
A great amount of research effort has been directed at platinum compounds that bind with DNA differently from cisplatin with the idea that the difference may translate into an altered spectrum of activity. Recently research has also been directed at applying combinations of platinum agents with tumour-active phytochemicals with the aim of providing a means of overcoming platinum resistance in ovarian cancer. Herein we report the synthesis of monofunctional platinum tris(3-hydroxypyridine)chloroplatinum(II) chloride (coded as LH1) and tris(imidazole)chloroplatinum(II) chloride (coded as LH2), and their activity alone and in combination with genistein and cisplatin against human ovarian A2780, cisplatin-resistant A2780(cisR) and picoplatin-resistant A2780(ZD0473R) cancer cell lines. Although both LH1 and LH2 were found to be less active than cisplatin against the tumour models, they produced synergistic outcomes in combination with genistein. Both the level of cellular accumulation of Pt and of Pt-DNA binding resulting from the combination were greater in the A2780(cisR) cell line than in the parental A2780 cell line, irrespective of the sequence of administration. Absence of association between activity of LH1 and LH2 and the level of Pt-DNA binding indicates that the cell death induced by LH1 and LH2 may not be limited to the effect of their binding with DNA.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether