  • Search Results
  • Pasteurella multocida toxin- induced osteoclastogenesis requires mTOR activation.
Pasteurella multocida toxin- induced osteoclastogenesis requires mTOR activation.
Cell communication and signaling : CCS (2015-09-16)
Bianca Kloos, Sushmita Chakraborty, Sonja G Lindner, Katrin Noack, Ulrike Harre, Georg Schett, Oliver H Krämer, Katharina F Kubatzky
ABSTRACT
Pasteurella multocida toxin (PMT) is a potent inducer of osteoclast formation. Pigs suffering from an infection with toxigenic Pasteurella multocida strains develop atrophic rhinitis characterised by a loss of turbinate bones and conchae. However, on the molecular level the process of bone loss remains largely uncharacterised. Recently it was found that PMT activates the serine/threonine kinase mammalian target of rapamycin (mTOR) in fibroblasts. Using RAW264.7 macrophages, we investigated the role of the mTOR complex 1 (mTORC1) in PMT-mediated osteoclast formation. PMT induces the differentiation of RAW264.7 macrophages into multinucleated, tartrate resistant acid phosphatase (TRAP) positive osteoclasts that are capable to resorb bone. In the presence of the mTORC1 inhibitor rapamycin, PMT was significantly less able to induce the formation of TRAP-positive osteoclasts. Accordingly, the resulting resorption of bone was strongly reduced. A major target of mTOR is the 70 kDa ribosomal protein S6 kinase 1 (p70 S6K1). Activated p70 S6K1 decreases the expression of programmed cell death protein 4 (PDCD4), a negative transcriptional regulator of osteoclastogenesis, at the protein and gene level. Ultimately this results in the activation of c-Jun, a component of the activator protein 1 (AP-1) complex, which is a major transcription factor for the induction of osteoclast-specific genes. We now demonstrate that c-Jun and its downstream target, the osteoclast-specific bone degrading protease cathepsin K, are upregulated upon PMT treatment in an mTOR-dependent manner. Activation of mTOR signalling plays a central role in the formation of osteoclasts through the bacterial toxin PMT. On the molecular level, PMT-induced activation of mTOR leads to down regulation of PDCD4, a known repressor of AP-1 complex, culminating in the activation of c-Jun, an essential transcription factor for triggering osteoclastogenesis.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%