  • Search Results
  • Chemical structure-related mechanisms underlying in vivo genotoxicity induced by nitrofurantoin and its constituent moieties in gpt delta rats.
Chemical structure-related mechanisms underlying in vivo genotoxicity induced by nitrofurantoin and its constituent moieties in gpt delta rats.
Toxicology (2015-03-17)
Aki Kijima, Yuji Ishii, Shinji Takasu, Kohei Matsushita, Ken Kuroda, Daisuke Hibi, Yuta Suzuki, Takehiko Nohmi, Takashi Umemura
ABSTRACT
Nitrofurans are antimicrobial compounds containing a nitro group at the 5-position of the furan ring and an amine or hydrazide side chain derivative. One member of the nitrofurans, nitrofurantoin (NFT), is a renal carcinogen in male rats despite its still controversial genotoxicity. We investigated chemical structure-related modes of action of NFT, and reporter gene mutation assays for NFT and its constituent moieties were performed. NFT, 5-nitro-2-furaldehyde (NFA), or 1-aminohydantoin (AHD) was administered to male F344 gpt delta rats by gavage for 4 or 13 weeks at a carcinogenic or the maximum tolerated dose. NFT caused a significant increase in gpt mutant frequency (MF) at 13 weeks with G-base substitution mutations. An increase in gpt MF was also observed in the NFA-treated group at 13 weeks, but not in the AHD-treated group. 8-Hydroxydeoxyguanosine (8-OHdG) levels in the kidney DNA of NFT-treated rats were significantly increased after 4 weeks. NFT caused accumulation of hyaline droplets indicated by positive immunostaining and western blot analysis for α2u-globulin in the proximal tubules. An additional study, in which female gpt delta rats were given NFT at the same dose used for males, was performed to mitigate the effect of α2u-globulin. NFT exerted the same effects on female rat kidneys to the same extent as males in terms of gpt MF and 8-OHdG level. Thus, it is highly probable that the structure of the nitro furan plays a key role in NFT-induced genotoxicity and genotoxic mechanisms including oxidative DNA damage are involved in NFT-induced renal carcinogenesis. α2u-globulin-mediated nephropathy may be a prerequisite for NFT-induced renal carcinogenesis in male rats, and additionally NFT could be a latent carcinogen in female rats and other animal species.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O