  • Search Results
  • Effects of insulin-like growth factor-1 on the relaxation responses of the cavernous smooth muscle from aged rats.
Effects of insulin-like growth factor-1 on the relaxation responses of the cavernous smooth muscle from aged rats.
Scandinavian journal of urology (2015-04-10)
Zhan-Ping Xu, Huai-Peng Wang, Jiu-Min Liu, Xiang-Guang Zheng, Dan Wu, Xiao-Yong Pu
ABSTRACT
The aim of this study was determine whether intracavernosal injection (ICI) of insulin-like growth factor-1 (IGF-1) protein can improve corpus cavernosal smooth muscle relaxation in aging rats. Ten young (4-month-old) and 30 old (24-month-old) Sprague-Dawley male rats were enrolled in the study. The old rats were divided into three groups: vehicle-only (n = 10), IGF-1 1 µg/kg (n = 10) and IGF-1 10 µg/kg treatment groups (n = 10). After 4 weeks of single IGF-1 injection treatment, strips of corporal tissue were precontracted with phenylephrine, and dose-response curves were generated to evaluate endothelial-dependent [acetylcholine (ACh)], endothelial-independent [sodium nitroprusside (SNP)] and electrical field stimulation (EFS) vasoreactivity. The changes in percentage of cavernosal smooth muscle and the concentration of nitric oxide (NO) in penile tissue were also evaluated. After IGF-1 treatment, the vasoreactivity was significantly improved in both the 1 µg/kg and the 10 µg/kg treatment groups compared with the vehicle-only group at 4 weeks in response to ACh, SNP and EFS (all p < 0.05). The percentage of cavernosal smooth muscle was increased in the IGF-1 treatment groups. The NO concentrations were increased after IGF-1 treatment. These data demonstrate that ICI of IGF-1 can improve vasoreactivity via endothelium-dependent and endothelial-independent mechanisms in the corpus cavernosum of the aging rat.
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon dioxide, ≥99.8%