  • Search Results
  • Phosphorylation of ULK1 by AMPK regulates translocation of ULK1 to mitochondria and mitophagy.
Phosphorylation of ULK1 by AMPK regulates translocation of ULK1 to mitochondria and mitophagy.
FEBS letters (2015-05-20)
Weili Tian, Wen Li, Yinqin Chen, Zeming Yan, Xia Huang, Haixia Zhuang, Wangtao Zhong, Yusen Chen, Wenxian Wu, Chunxia Lin, Hao Chen, Xiaoyan Hou, Liangqing Zhang, Senfang Sui, Bin Zhao, Zhe Hu, Longxuan Li, Du Feng
ABSTRACT
UNC-51 like kinase (ULK1) translocates to dysfunctional mitochondria and is involved in mitophagy, but the mechanisms responsible for ULK1 activation and translocation remain unclear. Here, we found that hypoxia induces phosphorylation of ULK1 at Serine-555 by Adenosine 5'-monophosphate (AMP)-activated protein kinase (AMPK). Unlike wild-type ULK1, an ULK1 (S555A) mutant cannot translocate to mitochondria in response to hypoxia. Inhibition or knockdown of AMPK prevents ULK1 translocation and inhibits mitophagy. Finally, the phospho-mimic ULK1 (S555D) mutant, but not ULK1 (S555A), rescues mitophagy in AMPK-knockdown cells. Thus, we conclude that AMPK-dependent phosphorylation of ULK1 is critical for translocation of ULK1 to mitochondria and for mitophagy in response to hypoxic stress.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride solution
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Anti-Atg1/ULK1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution