  • Search Results
  • Selenium Supplementation in Fish: A Combined Chemical and Biomolecular Study to Understand Sel-Plex Assimilation and Impact on Selenoproteome Expression in Rainbow Trout (Oncorhynchus mykiss).
Selenium Supplementation in Fish: A Combined Chemical and Biomolecular Study to Understand Sel-Plex Assimilation and Impact on Selenoproteome Expression in Rainbow Trout (Oncorhynchus mykiss).
PloS one (2015-05-16)
Davide Pacitti, Muhammad M Lawan, John Sweetman, Samuel A M Martin, Jörg Feldmann, Christopher J Secombes
ABSTRACT
Selenium (Se) is an essential oligonutrient, as a component of several Se-containing proteins (selenoproteins), which exert important biological functions within an organism. In livestock, Se-enriched products have been proposed as dietary supplements to be included into functional feeds for animal preventive health care. To this end, it is important to understand the optimal range of concentrations for supplementation and how long it takes to be assimilated into the organism. In this study, rainbow trout (Oncorhynchus mykiss) were fed a control diet containing 0.9 g Kg-1 Se or the same diet supplemented with a Se-Yeast product (Sel-Plex) to achieve Se concentrations ranging from 1.5-8.9 g Kg-1 for a period of ten weeks. Fish were sampled every two weeks for analysis. The kinetics of Se bioaccumulation and the effects on fish selenoprotein expression was determined in different tissues combining chemical and bimolecular techniques. The Sel-Plex enriched diets did not have any effect on survival and growth performance. The highest Se levels were found in liver and kidney followed by muscle and blood cells. Analysis of the Se concentration factor showed that liver is able to initially regulate the amount of Se accumulated. However, with higher dietary Se level (4.8 and 8.9 g Kg-1) and longer times of exposure (10 weeks), regulation is ineffective and the Se tissue concentration increases. The expression of the selected trout selenoprotein transcripts showed an inverse correlation with Sel-Plex augmentation in most cases. In liver, kidney and blood cells the highest up-regulation of the trout selenoprotein genes was seen mostly in the group fed the diet enriched with the lowest concentration of Sel-Plex (0.5 g Kg-1) for 10 weeks. Sel-Plex may represent an excellent Se supplement to deliver a high level of Se without provoking harm to the fish and to guarantee the maximal absorption of the element. According to our results, a dietary supplementation of Sel-Plex between 0.5 and 4 g Kg-1 may allow maximal benefits, whereas 8 g Kg-1 may be excessive for the purpose of supplementation.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade