  • Search Results
  • Development and validation of a method using supported liquid extraction for aldosterone determination in human plasma by LC-MS/MS.
Development and validation of a method using supported liquid extraction for aldosterone determination in human plasma by LC-MS/MS.
Clinica chimica acta; international journal of clinical chemistry (2015-05-20)
Cécile Meunier, Dominique Blondelle, Patrice Faure, Jean-Philippe Baguet, Caroline Le Goff, Olivier Chabre, Véronique Ducros
ABSTRACT
Accurate quantitation of aldosterone is essential for screening, diagnosis and subtype classification in primary aldosteronism. A simple, sensitive method for aldosterone in human plasma using supported liquid extraction (SLE) in combination with liquid chromatography tandem mass spectrometry (LC-MS/MS) was developed and validated. Plasma samples were diluted with water containing d7-aldosterone as internal standard. The samples were extracted with methyl-tert-butyl-ether (MTBE) on SLE cartridges. Separation was carried out on a Luna C18 (2) column using a methanol-water gradient. Detection was performed in the negative electrospray multiple reaction monitoring (MRM) quantitation. The use of water-based calibrators was evaluated against calibrators prepared in steroid-free serum. The assay was linear up to 3265pmol/L with an LOQ of approximately 40pmol/L. Within-run and between-run precision for plasma aldosterone were less than 10% except at low level near LOQ but were still less than 14.7% (Westgard's desirable specification). The mean recovery of the analyte added to plasma was greater than 97.7% and matrix effects were less than 4%. Comparison with another LC-MS/MS method was performed on a more sensitive instrument (ABSciex TQ 5500) and gave the equation API 3000=0.957×TQ 5500+12.6, linear regression r(2)=0.974 (n=43). An estimation of the reference interval for adults was established on a group of healthy volunteers (n=53). Calibration with water-based calibrators was validated and can be used for measurement of aldosterone by LC-MS/MS. This method is reliable, easy to perform on plasma specimens in a clinical environment and is attractive because of its simplicity.
Product Description
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Deuterium, 99.9 atom % D
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D