  • Search Results
  • A Transporter Motor Taken Apart: Flexibility in the Nucleotide Binding Domains of a Heterodimeric ABC Exporter.
A Transporter Motor Taken Apart: Flexibility in the Nucleotide Binding Domains of a Heterodimeric ABC Exporter.
Biochemistry (2015-05-08)
Magdalena A Bukowska, Michael Hohl, Eric R Geertsma, Lea M Hürlimann, Markus G Grütter, Markus A Seeger
ABSTRACT
ABC exporters are ubiquitous multidomain transport proteins that couple ATP hydrolysis at a pair of nucleotide binding domains to substrate transport across the lipid bilayer mediated by two transmembrane domains. Recently, the crystal structure of the heterodimeric ABC exporter TM287/288 was determined. One of its asymmetric ATP binding sites is called the degenerate site; it binds nucleotides tightly but is impaired in terms of ATP hydrolysis. Here we report the crystal structures of both isolated motor domains of TM287/288. Unexpectedly, structural elements constituting the degenerate ATP binding site are disordered in these crystals and become structured only in the context of the full-length transporter. In addition, hydrogen bonding patterns of key residues, including those of the catalytically important Walker B and the switch loop motifs, are fundamentally different in the solitary NBDs compared to those in the intact transport protein. The structures reveal crucial interdomain contacts that need to be established for the proper assembly of the functional transporter complex.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri