  • Search Results
  • Nuclear redox imbalance affects circadian oscillation in HaCaT keratinocytes.
Nuclear redox imbalance affects circadian oscillation in HaCaT keratinocytes.
The international journal of biochemistry & cell biology (2015-06-02)
Danilo Ranieri, Daniele Avitabile, Masaki Shiota, Akira Yokomizo, Seiji Naito, Mariano Bizzarri, Maria Rosaria Torrisi
ABSTRACT
Circadian clock is regulated by a transcriptional/translational feedback loop (TTFL) lasting ∼24 h. Circadian oscillation of peroxiredoxins (PRDX1-6) redox status has been shown in mature erythrocytes. We have recently reported that nuclear levels of PRDX2 are circadian regulated in the HaCaT keratinocytes. In this study, we addressed whether PRDX2 translocation could influence the TTFL. A reporter HaCaT cell line stably expressing the luciferase gene under control of Bmal1 promoter was lentivirally transduced either with an empty vector (EV), a vector carrying a myc-tagged wild type PRDX2 (PRDX2-Myc) or the same gene with a nuclear localization sequence (PRDX2-MycNuc). PRDX2 overexpressing cells were protected from H2O2-induced oxidative stress. The amplitude of the Bmal1 promoter activity was significantly dampened in PRDX2-MycNuc versus EV cells when synchronized either by dexamethasone treatment or temperature cycles. Clock synchronization was not affected in PRDX2 silenced cells. N-acetyl cysteine or melatonin treatments, significantly dampened the Bmal1 promoter activity suggesting that sustained scavenging of ROS impairs clock synchronization. Noteworthy, H2O2 treatment rescued proper oscillation of the clock in synchronized PRDX2-MycNuc HaCaT cells. Since the histone deacetylase Sirtuin 1 (Sirt1) modulates clock gene expression amplitude, the effect of Sirt1 activator resveratrol or Sirt1 inhibitor nicotinamide were also investigated. Interestingly, NAM enhanced the molecular clock synchronization in PRDX2-MycNuc cells. Our findings demonstrate that PRDX2 regulates the TTFL oscillation by finely tuning the cellular redox status of the nucleus likely influencing the deacetilase activity of SIRT1 enzyme.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
SAFC
HEPES
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)