  • Search Results
  • Comparative studies on exenatide-loaded poly (D,L-lactic-co-glycolic acid) microparticles prepared by a novel ultra-fine particle processing system and spray drying.
Comparative studies on exenatide-loaded poly (D,L-lactic-co-glycolic acid) microparticles prepared by a novel ultra-fine particle processing system and spray drying.
Colloids and surfaces. B, Biointerfaces (2015-06-04)
Chune Zhu, Ying Huang, Xiaoying Zhang, Liling Mei, Xin Pan, Ge Li, Chuanbin Wu
ABSTRACT
The purpose of this study was to compare the properties of exenatide-loaded poly (D,L-lactic-co-glycolic acid) microparticles (Ex-PLGA-MPs) prepared by a novel ultra-fine particle processing system (UPPS) and spray drying. UPPS is a proprietary technology developed by our group based on the disk rotation principle. Characteristics of the MPs including morphology, particle size distribution, drug content, encapsulation efficiency and in vitro release were comparatively studied. Cytotoxicity of the MPs was examined on A549 cells and the pharmacodynamics was investigated in vivo in type 2 diabetes Sprague-Dawley (SD) rats. Ex-PLGA-MPs prepared by UPPS showed larger particle size, denser surface, greater encapsulation efficiency, less initial burst release, and stable sustained release for more than one month in vitro as compared with the spray drying MPs. Meanwhile, the UPPS MPs effectively controlled the body growth rate and blood glucose in diabetes rats for at least three weeks after a single injection, while the spray drying MPs showed effective control period of about two weeks. UPPS technology was demonstrated to manufacture Ex-PLGA-MPs as a potential sustained release protein/polypeptide delivery system, which is an alternative method for the most commonly used spray drying. This comparative research provides a new guidance for microparticle preparation technology.
Sigma-Aldrich
Dimethyl carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl carbonate, anhydrous, ≥99%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)