Rotavirus disrupts cytoplasmic P bodies during infection.
Virus research (2015-09-20)
Rahul Bhowmick, Arpita Mukherjee, Upayan Patra, Mamta Chawla-Sarkar
ABSTRACT
Cytoplasmic Processing bodies (P bodies), the RNA-protein aggregation foci of translationally stalled and potentially decaying mRNA, have been reported to be differentially modulated by viruses. Rotavirus, the causative agent of acute infantile gastroenteritis is a double stranded RNA virus which completes its entire life-cycle exclusively in host cell cytoplasm. In this study, the fate of P bodies was investigated upon rotavirus infection. It was found that P bodies get disrupted during rotavirus infection. The disruption occurred by more than one different mechanism where deadenylating P body component Pan3 was degraded by rotavirus NSP1 and exonuclease XRN1 along with the decapping enzyme hDCP1a were relocalized from cytoplasm to nucleus. Overall the study highlights decay and subcellular relocalization of P body components as novel mechanisms by which rotavirus subverts cellular antiviral responses.
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
SAFC
HEPES
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, 0.2 μm filtered
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)