  • Search Results
  • Combined Experimental and Computational Studies of a Na2 Ni1-x Cux Fe(CN)6 Cathode with Tunable Potential for Aqueous Rechargeable Sodium-Ion Batteries.
Combined Experimental and Computational Studies of a Na2 Ni1-x Cux Fe(CN)6 Cathode with Tunable Potential for Aqueous Rechargeable Sodium-Ion Batteries.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-09-10)
Tai-Feng Hung, Hung-Lung Chou, Yu-Wen Yeh, Wen-Sheng Chang, Chang-Chung Yang
ABSTRACT
Herein, potential-tunable Na2 Ni1-x Cux Fe(CN)6 nanoparticles with three-dimensional frameworks and large interstitial spaces were synthesized as alternative cathode materials for aqueous sodium-ion batteries by controlling the molar ratio of Ni(II) to Cu(II) at ambient temperature. The influence of the value of x on the crystalline structure, lattice parameters, electrochemical properties, and charge transfer of the resultant compound was explored by using powder X-ray diffractometry, density functional theory, cyclic voltammetry, galvanostatic charge-discharge techniques, and Bader charge analysis. Of the various formulations investigated, that with x=0.25 delivered the highest reversible capacity, superior rate capability, and outstanding cycling performance. These attributes are ascribed to its unique face-centered cubic structure for facile sodium-ion insertion/extraction and the strong interactions between Cu and N atoms, which promote structural stability.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, puriss. p.a., ACS reagent, prima fine spirit, without additive, F15 o1, ~96%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, ReagentPlus®
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, BioReagent, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, puriss. p.a., ≥98%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)