  • Search Results
  • Roots extracts of Adenophora triphylla var. japonica improve obesity in 3T3-L1 adipocytes and high-fat diet-induced obese mice.
Roots extracts of Adenophora triphylla var. japonica improve obesity in 3T3-L1 adipocytes and high-fat diet-induced obese mice.
Asian Pacific journal of tropical medicine (2015-11-29)
Dong-Ryung Lee, Young-Sil Lee, Bong-Keun Choi, Hae Jin Lee, Sung-Bum Park, Tack-Man Kim, Han Jin Oh, Seung Hwan Yang, Joo-Won Suh
ABSTRACT
To investigate the anti-obesity activity and the action mechanism of the roots of Adenophora triphylla var. japonica extract (ATE) in high-fat diet (HFD)-induced obese mice and 3T3-L1 adipocytes. The roots of Adenophora triphylla were extracted with 70% ethanol. To demonstrate the compounds, linoleic acid was analyzed by using gas chromatography; and the anti-obesity effects and possible mechanisms of ATE were examined in 3T3-L1 adipocytes and HFD-induced obese mice. Treatment with ATE inhibited the lipid accumulation without cytotoxicity in 3T3-L1 adipocytes. Furthermore, 200 and 400 mg/kg ATE treatment significantly decreased the body weight gain, white adipose tissues (WATs) weight and plasma triglyceride level, while 100 and 200 mg/kg ATE treatment increased the plasma high-density lipoprotein cholesterol level in the HFD-induced obese mice, as compared with the HFD group. Treatment with 200 and 400 mg/kg ATE also lowered the size of adipocytes in adipose tissue and reduced the lipid accumulation in liver. ATE treatment showed significantly lower expression level of adipogenesis-related proteins, such as peroxisome proliferator-activated receptor γ, fatty acid binding protein (aP2), fatty acid synthase in 3T3-L1 adipocytes; and furthermore, decreased peroxisome proliferator-activated receptor γ, aP2, fatty acid synthase, sterol regulatory element binding protein-1c, and lipoprotein lipase mRNA expression levels in WAT of the HFD-induced obese mice. These results suggested that the ATE has an anti-obesity effect, which may be elicited by regulating the expression of adipogenesis and lipogenesis-related genes and proteins in adipocytes and WAT of the HFD-induced obese mice.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade