  • Search Results
  • Three-Layered Biodegradable Micelles Prepared by Two-Step Self-Assembly of PLA-PEI-PLA and PLA-PEG-PLA Triblock Copolymers as Efficient Gene Delivery System.
Three-Layered Biodegradable Micelles Prepared by Two-Step Self-Assembly of PLA-PEI-PLA and PLA-PEG-PLA Triblock Copolymers as Efficient Gene Delivery System.
Macromolecular bioscience (2015-02-04)
Daniel G Abebe, Rima Kandil, Teresa Kraus, Maha Elsayed, Olivia M Merkel, Tomoko Fujiwara
ABSTRACT
"Three-layered micelles" (3LM) composed of two triblock copolymers, poly(L-lactide)-b-polyethyleneimine-b-poly(L-lactide) (PLLA-PEI-PLLA) and poly(L-lactide)-b-poly(ethylene glycol)-b-poly(L-lactide) (PLLA-PEG-PLLA) are designed to combine electrostatic interaction and solvent-induced condensation of DNA. The low molecular weight PLLA-PEI-PLLA is synthesized by a facile amine-protection/deprotection approach and employed as a gene vector, compacting DNA as a polyplex core in the organo-micelles. The individual organo-micelle is further encapsulated within a PLLA-PEG-PLLA amphiphilic micelle leading to an aqueous stable colloidal dispersion. The resulting spherical 3LM possess a hydrodynamic diameter of ca. 200 nm and zeta potential close to neutral and display excellent stability to competing polyanions such as dextran sulfate in neutral pH (7.4). Such high stability is attributed to the complete shielding of the PEI/DNA polyplex core with an impermeable hydrophobic intermediate layer. However, greater than 90% of the encapsulated DNA are released within 30 min when exposed to slightly acidic pH (4.5). Based on our findings, a new class of non-viral delivery system for nucleic acids with superb stability and stealth properties is identified.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Tin(II) 2-ethylhexanoate, 92.5-100.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
1H-1,2,3-Triazole, 97%
Sigma-Aldrich
2-Methyl-2-oxazoline, 98%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.