  • Search Results
  • Effects of pinacidil on changes to the microenvironment around the incision site, of a skin/muscle incision and retraction, in a rat model of postoperative pain.
Effects of pinacidil on changes to the microenvironment around the incision site, of a skin/muscle incision and retraction, in a rat model of postoperative pain.
Molecular medicine reports (2015-03-12)
Su Cao, Yinbin Qin, Junjie Chen, Shiren Shen
ABSTRACT
The aim of the present study was to evaluate the influence of the microenvironment around an incision site, on peripheral and central sensitization. The effects of pinacidil activation of ATP-sensitive potassium (KATP) channels prior to skin/muscle incision and retraction (SMIR) surgery were assessed. A total of 24 male Sprague Dawley rats were randomly assigned to four groups: Control, sham (incision operation), SMIR (incision plus retraction 1 h after the skin/muscle incision) and pinacidil (SMIR plus pinacidil). The rats in the pinacidil group were intraperitoneally injected with pinacidil prior to the SMIR procedure. The mechanical withdrawal threshold (MWT) was determined at each time point. The microvessel density (MVD) value was determined by immunohistochemistry, and western blotting was performed to analyze the relative protein expression levels of nerve growth factor (NGF), glucose transporter protein-1 (GLUT1) and C-jun N-terminal kinases. There was a significant reduction in the levels of MVD, GLUT1 and MWT following SMIR surgery as compared with the incision alone, and a significant increase in the NGF protein expression levels. In the SMIR group, the MVD value was significantly increased seven days after surgery, as compared with three days after surgery. Additionally, intraperitoneal administration of pinacidil prior to the SMIR surgery inhibited the SMIR‑induced reduction in MWT and MVD and attenuated the SMIR‑induced GLUT1 reduction. The results of the present study suggest that the microenvironment around an incision site may affect the development of peripheral and central sensitization. In addition, pinacidil had an inhibitory effect on the formation of the inflammatory microenvironment around the incision site through activation of KATP channels, thereby inhibiting peripheral and central sensitization.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Picric acid, moistened with water, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Picric acid solution, 0.9-1.1% (alkalimetric)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri