  • Search Results
  • Exendin-4 protects rat islets against loss of viability and function induced by brain death.
Exendin-4 protects rat islets against loss of viability and function induced by brain death.
Molecular and cellular endocrinology (2015-05-16)
Rodrigo Carlessi, Natália E Lemos, Ana L Dias, Fernanda S Oliveira, Letícia A Brondani, Luis H Canani, Andrea C Bauer, Cristiane B Leitão, Daisy Crispim
ABSTRACT
Islet quality loss after isolation from brain-dead donors still hinders the implementation of human islet transplantation for treatment of type 1 diabetes. In this scenario, systemic inflammation elicited by donor brain death (BD) is among the main factors influencing islet viability and functional impairment. Exendin-4 is largely recognized to promote anti-inflammatory and cytoprotective effects on β-cells. Therefore, we hypothesized that administration of exendin-4 to brain-dead donors might improve islet survival and insulin secretory capabilities. Here, using a rat model of BD, we demonstrate that exendin-4 administration to the brain-dead donors increases both islet viability and glucose-stimulated insulin secretion. In this model, exendin-4 treatment produced a significant decrease in interleukin-1β expression in the pancreas. Furthermore, exendin-4 treatment increased the expression of superoxide dismutase-2 and prevented BD-induced elevation in uncoupling protein-2 expression. Such observations were accompanied by a reduction in gene expression of two genes often associated with endoplasmic reticulum (ER) stress response in freshly isolated islets from treated animals, C/EBP homologous protein and immunoglobulin heavy-chain binding protein. As ER stress response has been shown to be triggered by and to participate in cytokine-induced β-cell death, we suggest that exendin-4 might exert its beneficial effects through alleviation of pancreatic inflammation and oxidative stress, which in turn could prevent islet ER stress and β-cell death. Our findings might unveil a novel strategy to preserve islet quality from brain-dead donors. After testing in the human pancreatic islet transplantation setting, this approach might sum to the ongoing effort to achieve consistent and successful single-donor islet transplantation.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%