  • Search Results
  • Valproic acid exposure leads to upregulation and increased promoter histone acetylation of sepiapterin reductase in a serotonergic cell line.
Valproic acid exposure leads to upregulation and increased promoter histone acetylation of sepiapterin reductase in a serotonergic cell line.
Neuropharmacology (2015-07-08)
Diana Balasubramanian, Alicia X Deng, Kit Doudney, Mark B Hampton, Martin A Kennedy
ABSTRACT
Valproic acid (VPA) is a widely used antiepileptic drug and first-line treatment in bipolar disorder, although the mechanisms underlying its therapeutic effects are largely unknown. Recently, the recognition of VPA as an epigenetic drug offers new opportunities for understanding its therapeutic actions. In a rat serotonergic cell line (RN46A) we observed that VPA exposure has a strong upregulatory effect on the gene for sepiapterin reductase (SPR), a key enzyme involved in the tetrahydrobiopterin (BH4) synthetic pathway. BH4 is an essential cofactor in the biosynthesis of neurotransmitters like serotonin, dopamine and noradrenalin, and the BH4 pathway may thus be important in mood biology. Using real-time quantitative PCR we show that VPA, at therapeutically relevant doses, increases the expression of the Spr gene by about 8-fold in RN46A cells. In addition, Spr protein levels in VPA-exposed cells were elevated, as were the intracellular BH4 levels. HDAC inhibitors (HDACI) trichostatin A and sodium butyrate also upregulated Spr, but this was not observed using the VPA-analogue valpromide, which lacks HDAC inhibitory activity. Further examination of this effect revealed that exposure to VPA increased the acetylated histone mark H3K9/K14ac at the Spr promoter. The DNMT inhibitor 5'aza-dC also upregulated Spr by over 8-fold. However, DNA methylation status across the Spr promoter did not change in response to VPA. The BH4 pathway is fundamental to the regulation of neurotransmitters relevant to mood disorders, and this epigenetic effect of VPA at the Spr promoter may represent a novel mechanism through which VPA achieves its therapeutic action.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent