  • Search Results
  • Identification of Middle Chain Fatty Acyl-CoA Ligase Responsible for the Biosynthesis of 2-Alkylmalonyl-CoAs for Polyketide Extender Unit.
Identification of Middle Chain Fatty Acyl-CoA Ligase Responsible for the Biosynthesis of 2-Alkylmalonyl-CoAs for Polyketide Extender Unit.
The Journal of biological chemistry (2015-09-18)
Takeshi Miyazawa, Shunji Takahashi, Akihiro Kawata, Suresh Panthee, Teruo Hayashi, Takeshi Shimizu, Toshihiko Nogawa, Hiroyuki Osada
ABSTRACT
Understanding the biosynthetic mechanism of the atypical polyketide extender unit is important for the development of bioactive natural products. Reveromycin (RM) derivatives produced by Streptomyces sp. SN-593 possess several aliphatic extender units. Here, we studied the molecular basis of 2-alkylmalonyl-CoA formation by analyzing the revR and revS genes, which form a transcriptional unit with the revT gene, a crotonyl-CoA carboxylase/reductase homolog. We mainly focused on the uncharacterized adenylate-forming enzyme (RevS). revS gene disruption resulted in the reduction of all RM derivatives, whereas reintroduction of the gene restored the yield of RMs. Although RevS was classified in the fatty acyl-AMP ligase clade based on phylogenetic analysis, biochemical characterization revealed that the enzyme catalyzed the middle chain fatty acyl-CoA ligase (FACL) but not the fatty acyl-AMP ligase activity, suggesting the molecular evolution for acyl-CoA biosynthesis. Moreover, we examined the in vitro conversion of fatty acid into 2-alkylmalonyl-CoA using purified RevS and RevT. The coupling reaction showed efficient conversion of hexenoic acid into butylmalonyl-CoA. RevS efficiently catalyzed C8-C10 middle chain FACL activity; therefore, we speculated that the acyl-CoA precursor was truncated via β-oxidation and converted into (E)-2-enoyl-CoA, a RevT substrate. To determine whether the β-oxidation process is involved between the RevS and RevT reaction, we performed the feeding experiment using [1,2,3,4-(13)C]octanoic acid. (13)C NMR analysis clearly demonstrated incorporation of the [3,4-(13)C]octanoic acid moiety into the structure of RM-A. Our results provide insight into the role of uncharacterized RevS homologs that may catalyze middle chain FACL to produce a unique polyketide extender unit.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology