  • Search Results
  • Involvement of p38 MAPK activation mediated through AT1 receptors on spinal astrocytes and neurons in angiotensin II- and III-induced nociceptive behavior in mice.
Involvement of p38 MAPK activation mediated through AT1 receptors on spinal astrocytes and neurons in angiotensin II- and III-induced nociceptive behavior in mice.
Neuropharmacology (2015-07-26)
Wataru Nemoto, Yoshiki Ogata, Osamu Nakagawasai, Fukie Yaoita, Takeshi Tadano, Koichi Tan-No
ABSTRACT
We have previously demonstrated the possibility that angiotensin (Ang) II and its N-terminal metabolite Ang (1-7) act as neurotransmitters and/or neuromodulators in the spinal transmission of nociceptive information. Ang III, which is a C-terminal metabolite of Ang II, can also act on AT1 receptors, but its role in spinal nociceptive transmission remains unclear. Therefore, we examined the role of Ang III on the spinal nociceptive system in comparison with that of Ang II. Intrathecal (i.t.) administration of Ang III into mice produced a nociceptive behavior, which was dose-dependently inhibited by the co-administration of the AT1 receptor antagonist losartan and the p38 MAPK inhibitor SB203580, but not by the AT2 receptor antagonist PD123319, MEK1/2 inhibitor U0126 and JNK inhibitor SP600125. In addition, Ang III increased the phosphorylation of p38 MAPK in the dorsal lumbar spinal cord, which was inhibited by losartan. These effects were similar to those of observed with Ang II. The nociceptive behavior produced by Ang II or III was also attenuated by the administration of the astrocytic inhibitor L-α-aminoadipic acid, but not by the microglial inhibitor minocycline. Double immunohistochemical staining showed that spinal AT1 receptors were expressed on neurons and astrocytes, and that i.t. administration of either Ang II or III phosphorylated p38 MAPK in both spinal astrocytes and neurons. These results indicate that Ang III produces nociceptive behavior similar to Ang II, and suggest that the phosphorylation of p38 MAPK mediated through AT1 receptors on spinal astrocytes and neurons contributes to Ang II- and III-induced nociceptive behavior.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Bromophenol Blue
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)