  • Search Results
  • High doses of (-)-epigallocatechin-3-gallate from green tea induces cardiac fibrosis in mice.
High doses of (-)-epigallocatechin-3-gallate from green tea induces cardiac fibrosis in mice.
Biotechnology letters (2015-08-15)
Yi Cai, Shu-Qiao He, Hui-Qi Hong, Yue-Peng Cai, Li Zhao, Mei Zhang
ABSTRACT
Tea is the most consumed beverage in the world. (-)-Epigallocatechin-3-gallate (EGCG), a major green tea polyphenol, is effective in the prevention of several chronic diseases, and is marketed as part of many dietary supplements. We have now examined the myocardiotoxic effect of high doses of EGCG in mice. EGCG (500 and 1000 mg/kg·d) induced cardiac collagen synthesis and fibrosis-related protein expression, such as connective tissue growth factor (CTGF) and fibronectin (FN) in mice. Moreover, EGCG decreased the protein expression of p-AMPK and increased the levels of p-p70S6 K and p-S6. This is the first evidence that high oral doses of EGCG could induce cardiac fibrosis, and shed new light on the understanding of EGCG-mediated myocardiotoxicity.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea