  • Search Results
  • Functionalized graphene oxide-based thermosensitive hydrogel for magnetic hyperthermia therapy on tumors.
Functionalized graphene oxide-based thermosensitive hydrogel for magnetic hyperthermia therapy on tumors.
Nanotechnology (2015-08-21)
Xiali Zhu, Huijuan Zhang, Heqing Huang, Yingjie Zhang, Lin Hou, Zhenzhong Zhang
ABSTRACT
A novel locally injectable, biodegradable, and thermo-sensitive hydrogel made from chitosan and β-glycerophosphate salt was prepared. It incorporated polyethylenimine (PEI)-modified super-paramagnetic graphene oxide (GO/IONP/PEI) as a form of minimally invasive treatment of cancer lesions by magnetically induced local hyperthermia. Doxorubicin (DOX) was mixed into the hydrogel which was pre-loaded on GO/IONP/PEI to create a drug delivery system DOX-GO/IONP/PEI-gel. In addition to the evaluation of in vitro and in vivo antitumor activities, the physicochemical properties, magnetic properties and DOX release profile of the DOX-GO/IONP/PEI-gel were determined. The aqueous solution of the hydrogel showed a sol-gel transition behavior depending on temperature changes. Magnetization loops indicated the super-paramagnetic properties of GO/IONP/PEI. Compared with free DOX, DOX-GO/IONP/PEI could efficiently pass through cell membranes, leading to more apoptosis and demonstrating higher antitumor efficacy on MCF-7 cells in vitro. Furthermore, DOX-GO/IONP/PEI-gel intratumorally injected (i.t.) showed high antitumor efficacy on tumor-bearing mice in vivo, with no obvious toxicity. The antitumor efficacy was higher when combined with an alternating magnetic field (AMF), showing that DOX-GO/IONP/PEI-gel under AMF has great potential for cancer magnetic hyperthermia therapy.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
IR-783, Dye content 90 %
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)