  • Search Results
  • Okra (Hibiscus esculentus) gum-alginate blend mucoadhesive beads for controlled glibenclamide release.
Okra (Hibiscus esculentus) gum-alginate blend mucoadhesive beads for controlled glibenclamide release.
International journal of biological macromolecules (2014-10-15)
Priyanka Sinha, U Ubaidulla, Amit Kumar Nayak
ABSTRACT
The utility of isolated okra (Hibiscus esculentus) gum (OG) was evaluated as a potential sustained drug release polymer-blends with sodium alginate in the development of controlled glibenclamide release ionically-gelled beads for oral use. OG was isolated from okra fruits and its solubility, pH, viscosity and moisture content were studied. Glibenclamide-loaded OG-alginate blend beads were prepared using CaCl2 as cross-linking agent through ionic-gelation technique. These ionically gelled beads showed drug entrapment efficiency of 64.19 ± 2.02 to 91.86 ± 3.24%. The bead sizes were within 1.12 ± 0.11 to 1.28 ± 0.15 mm. These glibenclamide-loaded OG-alginate blend beads exhibited sustained in vitro drug release over a prolonged period of 8 h. The in vitro drug release from these OG-alginate beads were followed controlled-release (zero-order) pattern with super case-II transport mechanism. The beads were also characterized by SEM and FTIR. The swelling and degradation of these beads was influenced by the pH of the test medium. These beads also exhibited good mucoadhesivity with goat intestinal mucosa.
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis