  • Search Results
  • Long-term exposures to di-n-butyl phthalate inhibit body growth and impair gonad development in juvenile Murray rainbowfish (Melanotaenia fluviatilis).
Long-term exposures to di-n-butyl phthalate inhibit body growth and impair gonad development in juvenile Murray rainbowfish (Melanotaenia fluviatilis).
Journal of applied toxicology : JAT (2014-10-29)
Harpreet Bhatia, Anupama Kumar, John C Chapman, Mike J McLaughlin
ABSTRACT
The aim of the present study was to evaluate whether long-term exposures to environmentally relevant concentrations of di-n-butyl phthalate (DnBP) disrupt the reproduction-based endpoints in juvenile Murray rainbowfish (Melanotaenia fluviatilis). Fish were exposed to 5, 15 or 50 µg l(-1) DnBP for 30, 60 and 90 days each, and the effects on survival, body growth, whole-body concentrations of sex steroid hormones and gonadal development were investigated. The lowest observed effective concentration to affect the condition factor after 90 days was 5 µg l(-1). Complete feminization of the gonad was noted in fish exposed to 5 µg l(-1) for 90 days and to 15 and 50 µg l(-1) of DnBP for 30 or 60 days. After 90 days of exposure to DnBP, the ovaries were regressed and immature as opposed to the control fish which were in early-vitellogenic stage. Testes, present only in fish exposed to 5 µg l(-1) of DnBP for 30 or 60 days, were immature in comparison to the control fish that contained testes in the mid-spermatogenic phase. The E2/11-KT ratio was significantly higher only after exposures to 5 µg l(-1) DnBP for 90 days and 50 µg l(-1) DnBP for 30 days. Our data suggest that exposures to 5 µg l(-1) DnBP for 30 days did not have profound effects on body growth and gonadal differentiation of fish. However, 30 days of exposure to 15 µg l(-1) could interfere with the gonad development and to 50 µg l(-1) could compromise the hormonal profile of juvenile fish.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
11-Ketotestosterone