  • Search Results
  • Expression and purification recombinant antihypertensive peptide ameliorates hypertension in rats with spontaneous hypertension.
Expression and purification recombinant antihypertensive peptide ameliorates hypertension in rats with spontaneous hypertension.
Protein expression and purification (2015-05-13)
Xiao-Li Wang, Shi-Nan Ma, Ya-Hong Yuan, Yan Ding, Dong-Sheng Li
ABSTRACT
A highly efficient Escherichia coli expression system was established to obtain an appreciable quantity of antihypertensive peptide. The DNA-coding sequence for the Gly-Val-Tyr-Pro-His-Lys peptide was chemically synthesized and linked to form a ten-copy in tandem. It was cloned into the vector pET-15b and expressed in E. coli BL21 (DE3). The optimal conditions for maximal expression were verified and included the induction time and the concentration of isopropyl-β-D-thiogalactopyranoside. The recombinant protein was purified by affinity chromatography to greater than 95% purity, and further purification was achieved by High-performance Liquid Chromatography after cleavage with trypsin. The product was identified by Electrospray Ionization-Mass Spectrometry. The antihypertensive effects of the recombinant AHP were investigated in spontaneously hypertensive rats. The in vivo results demonstrated that a single oral administration of this peptide in spontaneously hypertensive rats resulted in a significant reduction of systolic blood pressure at 2h. Systolic blood pressure was stabilized 4h later and remained at a low level for 24h. This study provides a practical method to develop the peptide into functional foods or drugs for the prevention and treatment of hypertension.
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Sodium phosphate, 96%
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening