  • Search Results
  • Protective effects of butyrate on intestinal ischemia-reperfusion injury in rats.
Protective effects of butyrate on intestinal ischemia-reperfusion injury in rats.
The Journal of surgical research (2015-05-16)
Yingli Qiao, Jianmin Qian, Qingyang Lu, Yaqiang Tian, Qi Chen, Yang Zhang
ABSTRACT
Butyrate is normally fermented from undigested fiber by intestinal microflora. The goal of the present study was to determine the effects of butyrate and its underlying mechanisms on intestinal injury in a rat model of ischemia and reperfusion (I/R). Male Sprague-Dawley rats were subjected to warm ischemia for 45 min by clamping the superior mesenteric artery after treatment with butyrate, followed by 6 and 72 h of reperfusion. Pathologic histology analysis, enzyme-linked immunosorbent assay, immunofluorescence, and Western blot were performed. Butyrate preconditioning markedly improved intestinal injury. The inflammatory factor levels and leukocyte infiltration were attenuated by butyrate. Butyrate also maintained the intestinal barrier structures, increased the expression of tight junction proteins, and decreased endotoxin translocation. We conclude that butyrate administration attenuates intestinal I/R injury, which is associated with preservation of intestinal tight junction barrier function and suppression of inflammatory cell infiltration in the intestinal mucosa. This suggests butyrate as a potential strategy to prevent intestinal I/R injury.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
SAFC
Sodium butyrate
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)