  • Search Results
  • The Effect of Coumaric Acid on Ischemia-Reperfusion Injury of Sciatic Nerve in Rats.
The Effect of Coumaric Acid on Ischemia-Reperfusion Injury of Sciatic Nerve in Rats.
Inflammation (2015-06-08)
Mustafa Guven, Yasemin Yuksel, Muserref Hilal Sehitoglu, Mehmet Tokmak, Adem Bozkurt Aras, Tarik Akman, Umut Hatay Golge, Ferdi Goksel, Ergun Karavelioglu, Murat Cosar
ABSTRACT
The aim of the study was to determine the effect of coumaric acid on sciatic nerve ischemia/reperfusion (SNI) injury in rats. The rats were randomly divided into four groups: control group (no medication or surgical procedure), SNI group, SNI + coumaric acid (CA) group, and SNI + methylprednisolone (MP) group. Ischemia was achieved by abdominal aorta clamping, and all animals were sacrificed 24 h after ischemia. Harvested sciatic nerve segments were investigated histopathologically and for tissue biochemistry. A significant decrease in MDA, an increase in NRF1 levels, and increase in SOD activity were observed in the groups which received coumaric acid and methylprednisolone when compared to the corresponding untreated group (p < 0.05). Ischemic fiber degeneration significantly reduced in the SNI + CA and SNI + MP groups, especially in the SNI + MP group, compared to the SNI group (p < 0.05). Beta amyloid protein expressions were significantly decreased in the SNI + CA group compared to the SNI group (p < 0.05). Our study revealed that coumaric acid treatment after ischemia/reperfusion in rat sciatic nerves reduced oxidative stress and axonal degeneration. Therefore, coumaric acid may play a role in the treatment of peripheral nerve injuries due to ischemia/reperfusion.
Product Description
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.