  • Search Results
  • Analysis of polyunsaturated fatty acids and the omega-6 inflammatory pathway in hepatic ischemia/re-perfusion injury.
Analysis of polyunsaturated fatty acids and the omega-6 inflammatory pathway in hepatic ischemia/re-perfusion injury.
Molecular medicine reports (2015-06-13)
Ebru Kirac, Filiz Özcan, Hazal Tuzcu, Gulsum O Elpek, Mutay Aslan
ABSTRACT
The aim of the present study was to assess omega-3 (n-3) and omega-6 (n-6) polyunsaturated fatty acids (PUFAs) in liver tissue and evaluate changes in the n‑6-associated inflammatory pathway following liver ischemia/re‑perfusion (IR) injury. Male Wistar rats which were allowed free access to standard rat chow were included in the study. Blood vessels supplying the median and left lateral hepatic lobes were occluded with an arterial clamp for 60 min, followed by 60 min of re‑perfusion. Levels of arachidonic acid (AA, C20:4n‑6), dihomo‑gamma‑linolenic acid (DGLA, C20:3n‑6), eicosapentaenoic acid (EPA, C20:5n‑3) and docosahexaenoic acid (DHA, C22:6n‑3) in liver tissue were determined by an optimized multiple reaction monitoring method using ultra fast‑liquid chromatography coupled with tandem mass spectrometry. Phospholipase A2 (PLA2), cyclooxygenase (COX) and prostaglandin E2 (PGE2) were measured in tissue samples to evaluate changes in the n‑6 inflammatory pathway. Total histopathological score of cellular damage were significantly increased following hepatic IR injury. n‑3 and n‑6 PUFA levels were significantly increased in post‑ischemic liver tissue compared to those in non‑ischemic controls. No significant difference was observed in the AA/DHA and AA/EPA ratio in post‑ischemic liver tissues compared with that in the control. Tissue activity of PLA2 and COX as well as PGE2 levels were significantly increased in post‑ischemic liver tissues compared to those in non‑ischemic controls. The results of the present study suggested that increased hydrolysis of fatty acids via PLA2 triggers the activity of COX and leads to increased PGE2 levels. Future studies evaluating agents which block the formation of eicosanoids derived from n‑6 PUFAs may facilitate the development and application of treatment strategies in liver injury following IR.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Hematoxylin
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.