  • Search Results
  • Endothelial Cells Promote Pigmentation through Endothelin Receptor B Activation.
Endothelial Cells Promote Pigmentation through Endothelin Receptor B Activation.
The Journal of investigative dermatology (2015-08-27)
Claire Regazzetti, Gian Marco De Donatis, Houda Hammami Ghorbel, Nathalie Cardot-Leccia, Damien Ambrosetti, Philippe Bahadoran, Bérengère Chignon-Sicard, Jean-Philippe Lacour, Robert Ballotti, Andre Mahns, Thierry Passeron
ABSTRACT
Findings of increased vascularization in melasma lesions and hyperpigmentation in acquired bilateral telangiectatic macules suggested a link between pigmentation and vascularization. Using high-magnification digital epiluminescence dermatoscopy, laser confocal microscopy, and histological examination, we showed that benign vascular lesions of the skin have restricted but significant hyperpigmentation compared with the surrounding skin. We then studied the role of microvascular endothelial cells in regulating skin pigmentation using an in vitro co-culture model using endothelial cells and melanocytes. These experiments showed that endothelin 1 released by microvascular endothelial cells induces increased melanogenesis signaling, characterized by microphthalmia-associated transcription factor phosphorylation, and increased tyrosinase and dopachrome tautomerase levels. Immunostaining for endothelin 1 in vascular lesions confirmed the increased expression on the basal layer of the epidermis above small vessels compared with perilesional skin. Endothelin acts through the activation of endothelin receptor B and the mitogen-activated protein kinase, extracellular signal-regulated kinase (ERK)1/2, and p38, to induce melanogenesis. Finally, culturing of reconstructed skin with microvascular endothelial cells led to increased skin pigmentation that could be prevented by inhibiting EDNRB. Taken together these results demonstrated the role of underlying microvascularization in skin pigmentation, a finding that could open new fields of research for regulating physiological pigmentation and for treating pigmentation disorders such as melasma.
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
PTIO
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Tris(2-pyridylmethyl)amine, 98%