  • Search Results
  • Murine model of chemotherapy-induced extraintestinal pathogenic Escherichia coli translocation.
Murine model of chemotherapy-induced extraintestinal pathogenic Escherichia coli translocation.
Infection and immunity (2015-06-03)
Sabrina I Green, Nadim J Ajami, Li Ma, Nina M Poole, Roger E Price, Joseph F Petrosino, Anthony W Maresso
ABSTRACT
Escherichia coli is a major cause of life-threatening infections in patients with neutropenia, particularly those receiving chemotherapy for the treatment of cancer. In most cases, these infections originate from opportunistic strains living within the patient's gastrointestinal tract which then translocate to major organ systems. There are no animal models that faithfully recapitulate these infections, and, as such, the host or bacterial factors that govern this process remain unidentified. We present here a novel model of chemotherapy-induced bacterial translocation of E. coli. Oral gavage of BALB/c mice with a clinical isolate of extraintestinal pathogenic E. coli (ExPEC) leads to stable and long-term colonization of the murine intestine. Following the induction of neutropenia with the chemotherapeutic drug cyclophosphamide, ExPEC translocates from the intestine to the lungs, liver, spleen, and kidneys with concomitant morbidity in infected animals. Translocation can also occur in mice bearing mammary tumors, even in the absence of chemotherapy. Translocation of ExPEC is also associated with an increase of the diversity of bacterial DNA detected in the blood. This is the first report of a chemotherapy-based animal model of ExPEC translocation in cancerous mice, a system that can be readily used to identify important virulence factors for this process.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)