  • Search Results
  • Simultaneous Determination of Underivatized Vitamin B1 and B6 in Whole Blood by Reversed Phase Ultra High Performance Liquid Chromatography Tandem Mass Spectrometry.
Simultaneous Determination of Underivatized Vitamin B1 and B6 in Whole Blood by Reversed Phase Ultra High Performance Liquid Chromatography Tandem Mass Spectrometry.
PloS one (2015-07-03)
Johan Puts, Monique de Groot, Martin Haex, Bernadette Jakobs
ABSTRACT
Vitamin B1 (thiamine-diphosphate) and B6 (pyridoxal-5'phosphate) are micronutrients. Analysis of these micronutrients is important to diagnose potential deficiency which often occurs in elderly people due to malnutrition, in severe alcoholism and in gastrointestinal compromise due to bypass surgery or disease. Existing High Performance Liquid Chromatography (HPLC) based methods include the need for derivatization and long analysis time. We developed an Ultra High Performance Liquid Chromatography Tandem Mass spectrometry (UHPLC-MS/MS) assay with internal standards for simultaneous measurement of underivatized thiamine-diphosphate and pyridoxal-5'phosphate without use of ion pairing reagent. Whole blood, deproteinized with perchloric acid, containing deuterium labelled internal standards thiamine-diphosphate(thiazole-methyl-D3) and pyridoxal-5'phosphate(methyl-D3), was analyzed by UHPLC-MS/MS. The method was validated for imprecision, linearity, recovery and limit of quantification. Alternate (quantitative) method comparisons of the new versus currently used routine HPLC methods were established with Deming regression. Thiamine-diphosphate and pyridoxal-5'phosphate were measured within 2.5 minutes instrumental run time. Limits of detection were 2.8 nmol/L and 7.8 nmol/L for thiamine-diphosphate and pyridoxal-5'phosphate respectively. Limit of quantification was 9.4 nmol/L for thiamine-diphosphate and 25.9 nmol/L for pyridoxal-5'phosphate. The total imprecision ranged from 3.5-7.7% for thiamine-diphosphate (44-157 nmol/L) and 6.0-10.4% for pyridoxal-5'phosphate (30-130 nmol/L). Extraction recoveries were 101-102% ± 2.5% (thiamine-diphosphate) and 98-100% ± 5% (pyridoxal-5'phosphate). Deming regression yielded slopes of 0.926 and 0.990 in patient samples (n = 282) and national proficiency testing samples (n = 12) respectively, intercepts of +3.5 and +3 for thiamine-diphosphate (n = 282 and n = 12) and slopes of 1.04 and 0.84, intercepts of -2.9 and +20 for pyridoxal-5'phosphate (n = 376 and n = 12). The described UHPLC-MS/MS method allows simultaneous determination of underivatized thiamine-diphosphate and pyridoxal-5'phosphate in whole blood without intensive sample preparation.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Deuterium, 99.8 atom % D
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C