  • Search Results
  • An essential role of cAMP response element-binding protein in epidermal growth factor-mediated induction of sodium/glucose cotransporter 1 gene expression and intestinal glucose uptake.
An essential role of cAMP response element-binding protein in epidermal growth factor-mediated induction of sodium/glucose cotransporter 1 gene expression and intestinal glucose uptake.
The international journal of biochemistry & cell biology (2015-05-06)
Chun-Wen Wang, Wen-Liang Chang, Yu-Chuan Huang, Fang-Chi Chou, Fang-Na Chan, Shih-Chieh Su, Shu-Fen Huang, Hui-Hsuan Ko, Yi-Ling Ko, Hang-Chin Lin, Tsu-Chung Chang
ABSTRACT
The sodium/glucose cotransporter 1 (SGLT1) is responsible for glucose uptake in intestinal epithelial cells. Its expression is decreased in individuals with intestinal inflammatory disorders and is correlated with the pathogenesis of disease. The aim of this study was to understand the regulatory mechanism of the SGLT1 gene. Using the trinitrobenzene sulfonic acid-induced mouse models of intestinal inflammation, we observed decreased SGLT1 expression in the inflamed intestine was positively correlated with the mucosal level of epidermal growth factor (EGF) and activated CREB. Overexpression of EGF demonstrated that the effect of EGF on intestinal glucose uptake was primarily due to the increased level of SGLT1. We identified an essential cAMP binding element (CRE) confers EGF inducibility in the human SGLT1 gene promoter. ChIP assay further demonstrated the increased binding of CREB and CBP to the SGLT1 gene promoter in EGF-treated cells. In addition, the EGFR- and PI3K-dependent CREB phosphorylations are involved in the EGF-mediated SGLT1 expression. This is the first report to demonstrate that CREB is involved in EGF-mediated transcription regulation of SGLT1 gene in the normal and inflamed intestine, which can provide potential therapeutic applications for intestinal inflammatory disorders.
Sigma-Aldrich
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, 97%
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
4,4′-Bis(N-carbazolyl)-1,1′-biphenyl, sublimed grade, 99.9% trace metals basis
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)